Product Name
Atto 488 PPE suitable for fluorescence
Product Number
51028
Product Brand
SIGMA
CAS Number
Molecular Weight
Test
Specification
APPEARANCE (COLOR)Light Red to Dark Red
APPEARANCE (FORM)Powder or Crystals
PURITY (HPCE AREA %)≥ 90.0 %
SOLUBILITY (COLOR)REPORT RESULT
SOLUBILITY (TURBIDITY)Clear
SOLUBILITY (METHOD)10 nmol in 20 ml DMSO
EMISSION MAXIMUM527.0 ± 5.0 nm
REMARKS ON LUMINESCENCEEXC. 507 NM IN ETHANOL
RECOMMENDED RETEST PERIOD36 MONTHS