Product Name
Atto 633 DPPE suitable for fluorescence
Product Number
52816
Product Brand
SIGMA
CAS Number
Molecular Weight
Test
Specification
APPEARANCE (COLOR)Light Blue to Very Dark Blue
APPEARANCE (FORM)Powder or Crystals
PURITY (HPCE AREA %)≥ 90.0 %
SOLUBILITY (COLOR)REPORT RESULT
SOLUBILITY (TURBIDITY)Clear
SOLUBILITY (METHOD)10 nmol in 20 ml DMSO
EMISSION MAXIMUM655.0 ± 5.0 nm
REMARKS ON LUMINESCENCEEXC. 629 NM IN ETHANOL
RECOMMENDED RETEST PERIOD36 MONTHS