Product Name
Atto 465 NHS ester BioReagent, suitable for fluorescence, ≥90% (HPLC)
Product Number
53404
Product Brand
SIGMA
CAS Number
Molecular Weight
Test
Specification
APPEARANCE (COLOR)Report Result
APPEARANCE (FORM)Report Result
PURITY (HPLC AREA %)≥ 90 %
Absorption Max454 - 460 nm
SOLVENT (UV)acetonitrile with 0.1% acetic acid
DEGREE OF COUPLING≥ 90 %
RECOMMENDED RETEST PERIOD36 MONTHS