Product Name
6-Carboxytetramethylrhodamine BioReagent, suitable for fluorescence, ≥90% (HPLC)
Product Number
55222
Product Brand
SIGMA
CAS Number
91809-67-5
Molecular Weight
430.45
Test
Specification
APPEARANCE (COLOR)DEEP VIOLET OR DEEP GREEN TO BLACK
APPEARANCE (FORM)Powder
PURITY (HPLC AREA %)≥ 90.0 %
EMISSION MAXIMUM575 ± 5 nm
REMARKS ON LUMINESCENCEEX. 543 NM IN 0.1 M TRIS PH 8.0