Product Name
7-Hydroxy-N-octadecylcoumarin-3-carboxamide suitable for fluorescence, ≥98.0% (TLC)
Product Number
56059
Product Brand
SIGMA
CAS Number
Molecular Weight
457.65
Test
Specification
APPEARANCE (COLOR)White to Faint Yellow
APPEARANCE (FORM)Powder
PURITY (TLC AREA %)≥ 98.0 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
LUMINESCENCECORRESPONDS TO REQUIREMENTS
EMISSION MAXIMUM406.0 ± 5.0 nm