Product Name
Indigo carmine for microscopy (Bact., Hist.), indicator (pH 11.5-14.0)
Product Number
57000
Product Brand
SIGMA
CAS Number
860-22-0
Molecular Weight
466.35
Test
Specification
APPEARANCE (COLOR)Purple to Very Dark Purple
APPEARANCE (FORM)Powder
THIN LAYER CHROMATOGR.CORRESPONDS TO STANDARD
SOLUBILITY (COLOR)Blue to Very Dark Blue
SOLUBILITY (TURBIDITY)--
SOLUBILITY (METHOD)0.01G IN 10ML METHANOL
PH-TRANSITIONPH 11.5 = BLUE, PH 14.0 = YELLOW
LOSS ON DRYING≤ 10 %
WAVELENGTH (1) (UV)603 - 613 nm
EXTINCTION (1%/1CM) (1) (UV)≥ 350
SOLVENT (UV)Water