Product Name
Atto 594 iodoacetamide BioReagent, suitable for fluorescence
Product Number
59829
Product Brand
SIGMA
CAS Number
Molecular Weight
Test
Specification
APPEARANCE (COLOR)Light Red to Very Dark Red
APPEARANCE (FORM)Red to Very Dark Red
SOLUBILITY (COLOR)Blue to Very Dark Blue and Purple to Very Dark Purple
SOLUBILITY (TURBIDITY)Clear to Slightly Hazy
SOLUBILITY (METHOD)1 mg/ml in DMF
DEGREE OF COUPLING≥ 80.0 %
EMISSION MAXIMUM627 ± 10 nm
REMARKS ON LUMINESCENCEEx. 601 nm in 0.1 M phosphate pH 7.0
RECOMMENDED RETEST PERIOD36 MONTHS