Product Name
Kaempferol ≥97.0% (HPLC)
Product Number
60010
Product Brand
SIGMA
CAS Number
520-18-3
Molecular Weight
286.24
Test
Specification
APPEARANCE (COLOR)Faint Yellow to Yellow
APPEARANCE (FORM)Powder
PURITY (HPLC AREA %)≥ 97.0 %
WATER≤ 12 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE