Product Name
Atto 520 azide suitable for fluorescence
Product Number
61911
Product Brand
SIGMA
CAS Number
Molecular Weight
Test
Specification
APPEARANCE (COLOR)Light Red to Dark Red
APPEARANCE (FORM)Powder or Solid or Crystals
PURITY (HPCE AREA %)≥ 80.0 %
SOLUBILITY (COLOR)for information
SOLUBILITY (TURBIDITY)Clear
SOLUBILITY (METHOD)1 mg/ml in water
EMISSION MAXIMUM545 ± 10 nm
REMARKS ON LUMINESCENCEExc 516 nm in PBS, pH 7.4
RECOMMENDED RETEST PERIOD36 MONTHS