Product Name
DL-Methionine methylsulfonium chloride ≥97.0% (AT)
Product Number
64382
Product Brand
SIGMA
CAS Number
3493-12-7
Molecular Weight
199.70
Test
Specification
APPEARANCE (COLOR)Colorless or White
APPEARANCE (FORM)Powder
ARGENTOMETR. TITRATION97.0 - 103.0 %
SOLUBILITY (COLOR)Colorless
SOLUBILITY (TURBIDITY)Clear
SOLUBILITY (METHOD)1G IN 10ML WATER
PH4.0 - 5.5
LOSS ON DRYING≤ 3.0 %
RESIDUE ON IGNITION≤ 0.1 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE
MISCELLANEOUS TESTSHEAVY METALS ≤0.002 %