Product Name
7-Methoxycoumarin-3-carboxylic acid suitable for fluorescence, ≥97.0% (HPCE)
Product Number
64948
Product Brand
SIGMA
CAS Number
20300-59-8
Molecular Weight
220.18
Test
Specification
APPEARANCE (COLOR)White to Light Yellow
APPEARANCE (FORM)Powder
PURITY (HPCE AREA %)≥ 97.0 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EMISSION MAXIMUM402.0 ± 5.0 nm
REMARKS ON LUMINESCENCEEX. 330 NM IN 0.1 M TRIS PH 9.0