Product Name
Atto 390 azide BioReagent, suitable for fluorescence, ≥90% (HPLC)
Product Number
68321
Product Brand
SIGMA
CAS Number
Molecular Weight
543.66
Test
Specification
APPEARANCE (COLOR)Report Result
APPEARANCE (FORM)Report Result
PURITY (HPLC AREA %)≥ 90 %
Absorption Max380 - 384 nm
SOLVENT (UV)ethanol with 0.1% trifluoroacetic acid
RECOMMENDED RETEST PERIOD36 MONTHS