Product Name
Atto 532 maleimide BioReagent, suitable for fluorescence, ≥90% (coupling to thiols)
Product Number
68499
Product Brand
SIGMA
CAS Number
Molecular Weight
Test
Specification
APPEARANCE (COLOR)Dark Orange to Very Dark Orange and Dark Red Very Dark Red
APPEARANCE (FORM)Powder
PURITY (HPCE AREA %)≥ 90.0 %
SOLUBILITY (COLOR)FOR INFORMATION
SOLUBILITY (TURBIDITY)Clear
SOLUBILITY (METHOD)2 MG/ML IN DMF
DEGREE OF COUPLING≥ 90 %
EMISSION MAXIMUM553 ± 5 nm
REMARKS ON LUMINESCENCEEX. 532 NM IN 0.1 M PHOSPHATE PH 7.0