Product Name
Atto 633 azide suitable for fluorescence, ≥80.0% (HPCE)
Product Number
68517
Product Brand
SIGMA
CAS Number
Molecular Weight
Test
Specification
APPEARANCE (COLOR)Blue
APPEARANCE (FORM)Solid
PURITY (HPCE AREA %)≥ 80.0 %
SOLUBILITY (COLOR)Blue to Very Dark Blue
SOLUBILITY (TURBIDITY)Clear
SOLUBILITY (METHOD)1mg/ml in DMSO
EMISSION MAXIMUM657 ± 5 nm
REMARKS ON LUMINESCENCEEX. 629 NM IN 0.1 M PHOSPHATE PH 7.0
RECOMMENDED RETEST PERIOD36 MONTHS