Product Name
Atto 590 BioReagent, suitable for fluorescence, ≥90% (HPCE)
Product Number
70425
Product Brand
SIGMA
CAS Number
Molecular Weight
691.17
Test
Specification
APPEARANCE (COLOR)Purple to Dark Purple
APPEARANCE (FORM)Powder
PURITY (HPCE AREA %)≥ 90.0 %
SOLUBILITY (COLOR)VIOLET
SOLUBILITY (TURBIDITY)Clear
SOLUBILITY (METHOD)--
EMISSION MAXIMUM620 ± 10 nm
REMARKS ON LUMINESCENCEEX. 590 NM IN 0.1 M PHOSPHATE PH 7.0