Product Name
Sodium dodecyl sulfate ≥98.0% (GC)
Product Number
71729
Product Brand
SIGMA
CAS Number
151-21-3
Molecular Weight
288.38
Test
Specification
APPEARANCE (COLOR)Colorless or White
APPEARANCE (FORM)Powder or Powder with Chunk(s)
PURITY (GC AREA %)≥ 98.0 %
SOLUBILITY (COLOR)Colorless
SOLUBILITY (TURBIDITY)Clear
SOLUBILITY (METHOD)0.29 G IN 10 ML H2O (0.1 M)
LOSS ON DRYING≤ 5.0 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE
METAL TRACE ANALYSIS (ICP)CORRESPONDS TO REQUIREMENTS
CALCIUM (ICP)≤ 10 mg/kg
CADMIUM (ICP)≤ 5 mg/kg
COBALT (ICP)≤ 5 mg/kg
CHROMIUM (ICP)≤ 5 mg/kg
COPPER (ICP)≤ 5 mg/kg
IRON (ICP)≤ 5 mg/kg
POTASSIUM (ICP)≤ 200 mg/kg
MAGNESIUM (ICP)≤ 5 mg/kg
MANGANESE (ICP)≤ 5 mg/kg
NICKEL (ICP)≤ 5 mg/kg
LEAD (ICP)≤ 5 mg/kg
ZINC (ICP)≤ 5 mg/kg
CHLORIDE (CL)≤ 200 mg/kg