Product Name
Atto 565 NHS ester BioReagent, suitable for fluorescence, ≥70.0% (coupling to amines)
Product Number
72464
Product Brand
SIGMA
CAS Number
Molecular Weight
708.11
Test
Specification
APPEARANCE (COLOR)DEEP RED TO BLACK OR GREYISH-BLUE
APPEARANCE (FORM)Powder
SOLUBILITY (COLOR)FOR INFORMATION
SOLUBILITY (TURBIDITY)Clear
SOLUBILITY (METHOD)2 mg/ml in DMSO
DEGREE OF COUPLING≥ 70.0 %
EMISSION MAXIMUM590 ± 5 nm
REMARKS ON LUMINESCENCEEX. 565 NM IN 0.1 M PHOSPHATE PH 7.0
RECOMMENDED RETEST PERIOD28 MONTHS