Product Name
Atto 488 azide
Product Number
72709
Product Brand
SIGMA
CAS Number
Molecular Weight
Test
Specification
APPEARANCE (COLOR)REPORT RESULT
APPEARANCE (FORM)REPORT RESULT
EMISSION MAXIMUM523 ± 5 nm
REMARKS ON LUMINESCENCEIN 0.1 M PHOSPHATE PH 7.0