Product Name
Atto 655 iodoacetamide suitable for fluorescence
Product Number
73948
Product Brand
SIGMA
CAS Number
Molecular Weight
Test
Specification
APPEARANCE (COLOR)Blue to Very Dark Blue
APPEARANCE (FORM)Powder
SOLUBILITY (COLOR)Blue to Very Dark Blue
SOLUBILITY (TURBIDITY)Clear
EMISSION MAXIMUM684 ± 5 nm
REMARKS ON LUMINESCENCEEX. 663 NM IN 0.1 M PHOSPHATE PH 7.0
RECOMMENDED RETEST PERIOD36 MONTHS