Product Name
4-Methylumbelliferyl oleate suitable for fluorescence, ≥95% (HPCE)
Product Number
75164
Product Brand
SIGMA
CAS Number
18323-58-5
Molecular Weight
440.61
Test
Specification
APPEARANCE (COLOR)Colorless or White
APPEARANCE (FORM)Powder or Solid or Crystals or Crystalline Chunk(s) or Chunk(s)
PURITY (HPCE AREA %)≥ 95.0 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EMISSION MAXIMUM449 ± 5 nm
REMARKS ON LUMINESCENCEExc.: 327 nm in 0.1 M Phosphate, pH 7.0; Lipase
SUITABILITY ASSAYsuitable as a fluorogenic substrate for lipases