Product Name
Guanosine ≥97.0% (HPLC)
Product Number
75903
Product Brand
SIGMA
CAS Number
118-00-3
Molecular Weight
283.24
Test
Specification
APPEARANCE (COLOR)White to Faint Beige
APPEARANCE (FORM)Powder
PURITY (HPLC AREA %)≥ 97.0 %
SPECIFIC ROTATION (20/D)-73 ± 2 Degrees
CONCENTRATIONC=1.5 IN 0.1 M SODIUM HYDROXIDE
SOLUBILITY (COLOR)Colorless to Very Light Yellow
SOLUBILITY (TURBIDITY)Clear
SOLUBILITY (METHOD)1G IN 10ML 1 M SODIUM HYDROXIDE
WATER≤ 6.0 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE