Product Name
Atto 740 maleimide suitable for fluorescence
Product Number
77071
Product Brand
SIGMA
CAS Number
Molecular Weight
Test
Specification
APPEARANCE (COLOR)report result
APPEARANCE (FORM)Powder
SOLUBILITY (COLOR)Green to Very Dark Green
SOLUBILITY (TURBIDITY)Clear
SOLUBILITY (METHOD)1 MG/ML in DMF
DEGREE OF COUPLING≥ 80.0 %
EMISSION MAXIMUM764 ± 5 nm
REMARKS ON LUMINESCENCEEX. 740 NM IN 0.1 M PHOSPHATE PH 7.0