Product Name
Atto 520 NHS ester BioReagent, suitable for fluorescence, ≥80.0% (coupling to amines)
Product Number
77810
Product Brand
SIGMA
CAS Number
Molecular Weight
563.98
Test
Specification
APPEARANCE (COLOR)Red to Dark Red
APPEARANCE (FORM)Powder
SOLUBILITY (COLOR)FOR INFORMATION
SOLUBILITY (TURBIDITY)Clear
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
DEGREE OF COUPLING≥ 80.0 %
EMISSION MAXIMUM540 ± 5 nm
REMARKS ON LUMINESCENCEEX. 520 NM IN 0.1 M PHOSPHATE PH 7.0
RECOMMENDED RETEST PERIOD28 MONTHS