Product Name
Atto 610 BioReagent, suitable for fluorescence, ≥85.0% (HPCE/HPLC)
Product Number
78493
Product Brand
SIGMA
CAS Number
Molecular Weight
490.98
Test
Specification
PURITY (HPCE AREA %)≥ 85.0 %
LUMINESCENCECORRESPONDS TO REQUIREMENTS
EMISSION MAXIMUM635 ± 5 nm
REMARKS ON LUMINESCENCEnot specified