Product Name
Phenylmethanesulfonyl fluoride ≥99.0% (T)
Product Number
78830
Product Brand
SIGMA
CAS Number
329-98-6
Molecular Weight
174.19
Test
Specification
APPEARANCE (COLOR)Colorless or White
APPEARANCE (FORM)Powder or Crystals
TITRATION99.0 - 101.0 %
TITRATION (METHOD)-
PURITY (GC AREA %)≥ 99.0 %
MELTING POINT92 - 95 C
SOLUBILITY (COLOR)Colorless
SOLUBILITY (TURBIDITY)Clear
SOLUBILITY (METHOD)1G IN 10ML CHLOROFORM
WATER≤ 0.1 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE