Product Name
2-Ethylhexyl 4-methoxycinnamate 98%
Product Number
78848
Product Brand
SIGMA
CAS Number
5466-77-3
Molecular Weight
290.40
Test
Specification
APPEARANCE (COLOR)Colorless to Faint Yellow
APPEARANCE (FORM)Liquid or Viscous Liquid
PURITY (GC AREA %)≥ 97.5 %
REFRACTIVE INDEX N20/D1.542 - 1.548
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
RECOMMENDED RETEST PERIOD36 MONTHS