Product Name
Atto 550 PPE suitable for fluorescence
Product Number
78998
Product Brand
SIGMA
CAS Number
Molecular Weight
Test
Specification
APPEARANCE (COLOR)Light Red to Dark Red and Light Purple to Dark Purple
APPEARANCE (FORM)Powder or Crystals
PURITY (HPCE AREA %)≥ 90.0 %
SOLUBILITY (COLOR)REPORT RESULT
SOLUBILITY (TURBIDITY)Clear
SOLUBILITY (METHOD)10 nmol in 20 ml DMSO
EMISSION MAXIMUM578.0 ± 5.0 nm
REMARKS ON LUMINESCENCEEX. 556 NM IN ETHANOL
RECOMMENDED RETEST PERIOD36 MONTHS