Product Name
Rhodamine 101 inner salt for fluorescence
Product Number
83694
Product Brand
SIGMA
CAS Number
116450-56-7
Molecular Weight
490.59
Test
Specification
APPEARANCE (COLOR)Purple to Very Dark Purple
APPEARANCE (FORM)Powder or Crystals
WAVELENGTH (1) (UV)562.0 - 572.0 nm
EXTINCTION (1%/1CM) (1) (UV)≥ 1600
SOLVENT (UV)methanol
EMISSION MAXIMUM589 ± 5 NM
REMARKS ON LUMINESCENCEEXC.: 560 NM IN METHANOL