Product Name
D-Ribulose 1,5-bisphosphate sodium salt hydrate ≥99.0% (TLC)
Product Number
83895
Product Brand
SIGMA
CAS Number
24218-00-6
Molecular Weight
310.09 (anhydrous free acid basis)
Test
Specification
APPEARANCE (COLOR)White to Off White
APPEARANCE (FORM)Powder
PURITY (TLC AREA %)≥ 99.0 %
SOLUBILITY (COLOR)Colorless to Light Yellow
SOLUBILITY (TURBIDITY)Clear
SOLUBILITY (METHOD)50MG/ML WATER
WATER≤ 16 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE