Product Name
Blue Dextran average mol wt 10,000
Product Number
90475
Product Brand
SIGMA
CAS Number
87915-38-6
Molecular Weight
Test
Specification
APPEARANCE (COLOR)Blue to Dark Blue
APPEARANCE (FORM)Powder
MOLECULAR NUMBERNUMBER AVERAGE MOLECULAR WEIGHT (MN) REPORT RESULT
MOLECULAR WEIGHT DETERMINATIONWEIGHT AVERAGE MOLECULAR WEIGHT (MW) 9000 - 12000
LOSS ON DRYING≤ 10 %
MISCELLANEOUS TESTSLOADING 0.01 - 0.08 MMOL/G