Product Name
Atto 740 BioReagent, suitable for fluorescence, ≥98.0% (HPCE)
Product Number
91394
Product Brand
SIGMA
CAS Number
Molecular Weight
Test
Specification
APPEARANCE (COLOR)Light Blue to Dark Blue and Light Green to Dark Green
APPEARANCE (FORM)Powder
PURITY (HPCE AREA %)≥ 98.0 %
SOLUBILITY (COLOR)FOR INFORMATION
SOLUBILITY (TURBIDITY)Clear
SOLUBILITY (METHOD)1 MG/ML IN DMF
EMISSION MAXIMUM764 ± 5 nm
REMARKS ON LUMINESCENCEEX. 740 NM IN 0.1 M PHOSPHATE PH 7.0