Product Name
Atto 565-Biotin BioReagent, suitable for fluorescence, ≥90% (HPCE)
Product Number
92637
Product Brand
SIGMA
CAS Number
Molecular Weight
921.50
Test
Specification
APPEARANCE (COLOR)DEEP VIOLET TO BLACK-BLUE
APPEARANCE (FORM)Powder
PURITY (HPCE AREA %)≥ 90.0 %
SOLUBILITY (COLOR)DEEP RED TO DEEP VIOLET
SOLUBILITY (TURBIDITY)Clear
SOLUBILITY (METHOD)2 MG/ML IN DMF
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EMISSION MAXIMUM590 ± 5 nm
REMARKS ON LUMINESCENCEEX. 565 NM IN 0.1 M PHOSPHATE PH 7.0