Product Name
Atto 620 BioReagent, suitable for fluorescence, ≥90% (HPCE)
Product Number
92716
Product Brand
SIGMA
CAS Number
Molecular Weight
Test
Specification
APPEARANCE (COLOR)DEEP BLUE TO DEEP VIOLET
APPEARANCE (FORM)Powder
PURITY (HPCE AREA %)≥ 90.0 %
SOLUBILITY (COLOR)Blue to Very Dark Blue
SOLUBILITY (TURBIDITY)Clear
SOLUBILITY (METHOD)1 MG/ML IN DMSO
EMISSION MAXIMUM641 ± 5 nm
REMARKS ON LUMINESCENCEEx.: 617 nm in 0.1 M phosphate pH 7.0