Product Name
5-Carboxyfluorescein N-succinimidyl ester BioReagent, suitable for fluorescence, ≥90% (HPLC)
Product Number
92846
Product Brand
SIGMA
CAS Number
92557-80-7
Molecular Weight
473.39
Test
Specification
APPEARANCE (COLOR)Yellow to Dark Yellow and Orange to Dark Orange
APPEARANCE (FORM)Powder
PURITY (HPLC AREA %)≥ 90 %
LUMINESCENCECORRESPONDS TO REQUIREMENTS
DEGREE OF COUPLING≥ 90.0 %
EMISSION MAXIMUM515 - 525 nm
RECOMMENDED RETEST PERIOD42 MONTHS