Product Name
Atto 514 amine suitable for fluorescence
Product Number
93728
Product Brand
SIGMA
CAS Number
Molecular Weight
Test
Specification
APPEARANCE (COLOR)Light Yellow to Very Dark Yellow and Light Brown-Yellow to Very Dark Brown-Yellow
APPEARANCE (FORM)Powder or Solid
SOLUBILITY (COLOR)for information
SOLUBILITY (TURBIDITY)Clear
SOLUBILITY (METHOD)1 mg/ml in DMF
EMISSION MAXIMUM533 ± 10 nm
REMARKS ON LUMINESCENCEEx. 511 nm in PBS pH 7.4
RECOMMENDED RETEST PERIOD36 MONTHS