Product Name
Atto 647 BioReagent, suitable for fluorescence, ≥90% (HPCE)
Product Number
97875
Product Brand
SIGMA
CAS Number
Molecular Weight
Test
Specification
APPEARANCE (COLOR)Blue to Very Dark Blue and Blue-Green
APPEARANCE (FORM)Powder
PURITY (HPCE AREA %)≥ 90.0 %
SOLUBILITY (COLOR)BLUE TO TURQUOISE
SOLUBILITY (TURBIDITY)Clear
SOLUBILITY (METHOD)2 mg/ml in DMF
EMISSION MAXIMUM665 ± 5 nm
REMARKS ON LUMINESCENCEEX. 643 NM IN 0.1 M PHOSPHATE PH 7.0