Product Name
Ala-Gly
Product Number
A0878
Product Brand
SIGMA
CAS Number
687-69-4
Molecular Weight
146.14
Test
Specification
APPEARANCEWHITE POWDER
SOLUBILITYCLEAR TO SLIGHTLY HAZY COLORLESS SOLUTION AT 50 MG/ML IN 80% ACETIC ACID
CARBON39.5% TO 42.0%
NITROGEN18.5% TO 20.0%
SPECIFIC ROTATION+49.5 DEG TO 51.5 (C = 2.5 IN WATER AT 24 DEG C)
PURITY BY THIN LAYER CHROMATOGRAPHY> OR = 99%
DOCUMENT #A0878/11/06/07/1