Product Name
8-(6-Aminohexyl)aminoadenosine 3′:5′-cyclic monophosphate ~98%
Product Number
A8138
Product Brand
SIGMA
CAS Number
39824-30-1
Molecular Weight
443.39
Test
Specification
APPEARANCEWHITE POWDER
SOLUBILITYCLEAR COLORLESS SOLUTION AT 5 MG PLUS 0.1 ML OF WATER
WATER CONTENT BY KARL FISCHER< OR = 12.0%
UV-VIS SPECTRUMEMM = 14.5 TO 15.7 AT LAMBDA MAX 275 NM TO 277 NM, 0.1M HCL (ANHYDROUS)
PURITY BY HPLC> OR = 97%
DOCUMENT #A8138/01/12/09/1