Product Name
Anti-AARS antibody produced in rabbit affinity isolated antibody
Product Number
AV40369
Product Brand
SIGMA
CAS Number
Molecular Weight
Test
Specification
Concentration0.5-1 mg/ml
ELISANot Verified
Western BlottingAntibody dilution: 1 µg/ml
ImmunohistochemistryNot Verified
ImmunofluorescenceNot Verified
ImmunoprecipitationNot Verified
Flow CytometryNot Verified
Chromatin ImmunoprecipitationNot Verified