Product Name
5-Bromo-4-chloro-3-indolyl butyrate ≥99%
Product Number
B9151
Product Brand
SIGMA
CAS Number
129541-43-1
Molecular Weight
316.58
Test
Specification
APPEARANCE (COLOR)White
APPEARANCE (FORM)Powder
PURITY (TLC AREA %)≥ 98.5 %
SOLUBILITY (COLOR)Colorless
SOLUBILITY (TURBIDITY)Clear
SOLUBILITY (METHOD)5MG PLUS 0.1ML OF ACETONE
INFRARED SPECTRUMCONFORMS TO STRUCTURE