Product Name
Cytidine 5′-triphosphate disodium salt ≥95%
Product Number
C1506
Product Brand
SIGMA
CAS Number
36051-68-0
Molecular Weight
527.12
Test
Specification
APPEARANCE (COLOR)Colorless or White
APPEARANCE (FORM)Powder
PURITY (HPLC AREA %)≥ 95 %
SOLUBILITY (COLOR)Colorless
SOLUBILITY (TURBIDITY)Clear
SOLUBILITY (METHOD)50 MG/ML IN WATER
WATER≤ 12.0 %
WAVELENGTH (1) (UV)271.0 - 272.0 nm
MOLAR ABSORBANCY INDEX (1)EMM = 8.5 - 9.5 (water and solvent free basis)
SOLVENT (UV)0.1 M PHOSPHATE BUFFER PH 7.0
RESIDUAL SOLVENTS (GLC-HS)≤ 4 %