Product Name
Cortisone ≥98%
Product Number
C2755
Product Brand
SIGMA
CAS Number
53-06-5
Molecular Weight
360.44
Test
Specification
APPEARANCE (COLOR)White to Off White
APPEARANCE (FORM)Powder
PURITY (HPLC AREA %)≥ 98 %
SOLUBILITY (COLOR)Colorless to Faint Yellow
SOLUBILITY (TURBIDITY)Clear to Very Slightly Hazy
SOLUBILITY (METHOD)200MG PLUS 4ML OF CHLOROFORM:METHANOL (1:1)
INFRARED SPECTRUMCONFORMS TO STRUCTURE
WAVELENGTH (1) (UV)LAMBDA MAX 238-239 NM
MOLAR ABSORBANCY INDEX (1)EMM = 15.4-16.0
SOLVENT (UV)Methanol