Product Name
Collagen from mouse sternum Bornstein and Traub Type II, acid soluble, powder
Product Number
C5733
Product Brand
SIGMA
CAS Number
9007-34-5
Molecular Weight
Test
Specification
SolubilityConforms
 Soluble in 0.01 M to 0.5 M HOAc
Proteoglycan Content≤0.4 %