Product Name
Chloroform ≥99%, PCR Reagent, contains amylenes as stabilizer
Product Number
C7559
Product Brand
SIGMA
CAS Number
67-66-3
Molecular Weight
119.38
Test
Specification
Appearance (Color)Colorless
Appearance (Form)Liquid
Infrared spectrumConforms to Structure
Purity (GC)≥99.00 %
DNAse, Exonuclease DetectionNone Detected
NICKase,Endonuclease DetectionNone Detected
RNAse DetectionNone Detected