Product Name
Protein Kinase A catalytic subunit human >90% (SDS-PAGE), recombinant, expressed in E. coli, buffered aqueous glycerol solution
Product Number
C8482
Product Brand
SIGMA
CAS Number
Molecular Weight
Test
Specification
mg protein/ml (Bradford)≥0.1
units/mg protein≥1000
Unit Definition-------------------------
 One unit will transfer 1 nanomole of phosphate
 from 32P-ATP to kemptide substrate per minute at
 pH 7.4 at 30 deg C.
SDS ElectrophoresisConforms
 (Purity), > or = 90%