Product Name
Dulbecco’s Modified Eagle’s Medium - low glucose 10 ×, With 1000 mg/L glucose (1x), without L-glutamine, sodium bicarbonate, and folic acid, liquid, sterile-filtered, suitable for cell culture
Product Number
D2429
Product Brand
SIGMA
CAS Number
Molecular Weight
Test
Specification
Appearance (Turbidity)Clear
Appearance (Form)Solution
pH1.8 - 2.0
Cell LineCell Line - Cell Types
SterilityPass
Endotoxin at 1X<= 1 EU/ml
Glucose Concentration9.0 - 11.0 g/l