Product Name
Dansyl chloride BioReagent, suitable for amino acid labeling, powder and chunks, ≥99% (HPLC)
Product Number
D2625
Product Brand
SIGMA
CAS Number
605-65-2
Molecular Weight
269.75
Test
Specification
APPEARANCEYELLOW TO YELLOW-ORANGE POWDER AND CHUNKS
SOLUBILITYCLEAR TO VERY HAZY YELLOW TO VERY DEEP YELLOW-ORANGE SOLUTION AT 20MG/ML IN ACETONE
ELEMENTAL ANALYSIS52.4 TO 54.4% CARBON
4.9 TO 5.5% NITROGEN
PURITY BY HPLC> OR = 99%
SUITABILITY TESTSUITABLE FOR DANSYLATION OF AMINO ACIDS.
DOCUMENT # D2625/12/19/07/4