Product Name
Dimethyl sulfoxide Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Product Number
D2650
Product Brand
SIGMA
CAS Number
67-68-5
Molecular Weight
78.13
Test
Specification
Appearance (Turbidity)Clear
Appearance (Colour)Colorless
Appearance (Form)Liquid
SterilityPass
Endotoxin Level<= 1 EU/ml
Cytoxicity or abnormal cell growth (CF)Not Detected
Purity (GC)>= 99.7 %
Water Content (From Raw Material)<= 0.1 %
ID by IR. NIR or NMR (From Raw Material)Pass
Animal Derived Component FreeConforms