Product Name
1,2-Dioctanoyl-sn-glycerol ≥96% (TLC), oil
Product Number
D5156
Product Brand
SIGMA
CAS Number
60514-48-9
Molecular Weight
344.49
Test
Specification
Appearance (Turbidity)Clear
Appearance (Color)Colorless
Appearance (Form)Liquid
Solubility (Color)Colorless
Solubility (Turbidity)Clear
 25 mg + 0.5 mL, CHCl3
Proton NMR spectraConforms to Structure
Purity by Gas Chromatography≥99 %
 (as octanoic acid)
Purity (TLC)≥96 %
Recommended Retest Period------------------------------
 3 years