Product Name
MISSION® esiRNA targeting human KIFC3
Product Number
EHU024561
Product Brand
SIGMA
CAS Number
Molecular Weight
Test
Specification
AppearanceColorless Liquid
Concentration200 ng/µl
PCR Product - Gel ElectrophoresisPass
PCR Product - DNA SequencingPass
Concentration - OD260 or Fluorescence IntensityPass
Purified RNaseIII - Gel ElectrophoresisPass